missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZIM3 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA556979

Product Code. 15724881

  • € 566.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: NYSNLVSVGQ GETTKPDVIL RLEQGKEPWL EEEEVLGSGR AEKNGDIGGQ IWKPKDVKES LAREVPSINK ETLTTQKGVE CDGSKK Highest antigen sequence identity to the following orthologs: Mouse - 33%, Rat - 35%.

ZIM3 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 11 C2H2-type zinc fingers and 1 KRAB domain. ZIM3 may be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ZIM3
Polyclonal
Unconjugated
ZIM3
ZIM3; Zinc finger imprinted 3; zinc finger protein 657; zinc finger, imprinted 3; ZNF657
Rabbit
Antigen affinity chromatography
RUO
114026
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q96PE6
ZIM3
Recombinant protein corresponding to Human ZIM3. Recombinant protein control fragment (Product #RP-95514).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.