missing translation for 'onlineSavingsMsg'
Learn More

Zinc Diethyldithiocarbamate, TRC

Product Code. 30011676
Change view
Click to view available options
Quantity:
25 g
50 g
Unit Size:
25g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011676 25 g 25g
30005812 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011676 Supplier Toronto Research Chemicals Supplier No. TRCZ43999825G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14324-55-1
Formula Weight 359.9801
InChI Formula InChI=1S/2C5H11NS2.Zn/c2*1-3-6(4-2)5(7)8;/h2*3-4H2,1-2H3,(H,7,8);/q;;+2/p-2
IUPAC Name zinc;N,N-diethylcarbamodithioate
Molecular Formula C10H20N2S4Zn
Molecular Weight (g/mol) 361.92
SMILES [Zn+2].CCN(CC)C(=S)[S-].CCN(CC)C(=S)[S-]
Synonym Bis(N,N-diethyldithiocarbamato)zinc, Bis(diethyldithiocarbamato)zinc, Bis(diethyldithiocarbamato)zinc(II), Zinc N,N-diethyldithiocarbamate, Zinc Bis(N,N-diethyldithiocarbamate), Zinc bis(diethyldithiocarbamate), Zinc Diethylcarbamodithioate
Chemical Name or Material Zinc Diethyldithiocarbamate

Specifications

Quantity 25 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.