missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc finger and BTB domain-containing protein 9 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76522AF594
This item is not returnable.
View return policy
Description
Zinc finger and BTB domain-containing protein 9 Polyclonal antibody specifically detects Zinc finger and BTB domain-containing protein 9 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Zinc finger and BTB domain-containing protein 9 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 594 | |
| MGC23166, zinc finger and BTB domain containing 9, zinc finger and BTB domain-containing protein 9, ZNF919 | |
| Antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human ZBTB9. The immunogen is located within the first 50 amino acids of ZBTB9. Amino Acid Squence: PQHSSSLLESLNRHR | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| 221504 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction