missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc finger protein 180 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP1-92612
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Descrizione
Zinc finger protein 180 Polyclonal specifically detects Zinc finger protein 180 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifica
| Zinc finger protein 180 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| HHZ168zinc finger protein 180 (HHZ168), zinc finger protein 180 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 7733 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| ZNF180 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:EESMEEQDEKPPEPPKACAQDSFLPQEIIIKVEGEDTGSLTIPSQEGVNFKIVTVDFTREEQGTWNPAQRTLDRDVILENHRDLVSWDLATAVGKKDSTSKQRIFDEEPANGVKIERFTRDD | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto