missing translation for 'onlineSavingsMsg'
Learn More

Hach™ ZincoVer™ 5 Zinc Reagent Powder Pillows

Product Code. 10400694
Click to view available options
Range:
0.01 to 2.00mg/L Zn
0.01 to 3.00mg/L Zn
Sample Size:
20 mL
50 mL
No. of Tests:
25
100
Unit Size:
Each
Pack of 25
This item is not returnable. View return policy

Product Code. 10400694

Brand: Hach™ 2106669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ready-to-use, premeasured powder reagent in individual sealed packages for determination of zinc by the ZincoVer™ 5 zincon method. US EPA approved for wastewater analysis.

Powder Pillows make titration applications quick and easy.

  • Sealed packets ensure fresh, uncontaminated reagent every time
  • Premeasured to avoid measurement error and contamination from handling
  • Ready to use packets save time and hassle
  • Exact doses to easily monitor supply and prepare for lab procedures
  • Single-use packs for easy transportation and packing in test kits

Specifications

Analytes Zinc
Certifications/Compliance US EPA
Format Individual Packet
For Use With (Application) Zinc determination
No. of Tests 100
Quantity 100 Pack
Range 0.01 to 3.00mg/L Zn
Sample Size 20 mL
Test Method Zincon
Type Reagent packet
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.