Learn More
Hach™ ZincoVer™ 5 Zinc Reagent Powder Pillows
Click to view available options
Range:
0.01 to 2.00mg/L Zn
0.01 to 3.00mg/L Zn
Sample Size:
20 mL
50 mL
No. of Tests:
25
100
Unit Size:
Each
Pack of 25
Description
Powder Pillows make titration applications quick and easy.
- Sealed packets ensure fresh, uncontaminated reagent every time
- Premeasured to avoid measurement error and contamination from handling
- Ready to use packets save time and hassle
- Exact doses to easily monitor supply and prepare for lab procedures
- Single-use packs for easy transportation and packing in test kits
Specifications
Specifications
| Analytes | Zinc |
| Certifications/Compliance | US EPA |
| Format | Individual Packet |
| For Use With (Application) | Zinc determination |
| No. of Tests | 100 |
| Quantity | 100 Pack |
| Range | 0.01 to 3.00mg/L Zn |
| Sample Size | 20 mL |
| Test Method | Zincon |
| Type | Reagent packet |