Learn More
Hach™ ZincoVer™ 5 Zinc Reagent Powder Pillows
Ready-to-use, premeasured powder reagent in individual sealed packages for determination of zinc by the ZincoVer™ 5 zincon method. US EPA approved for wastewater analysis.
Brand:  Hach™ 1403268
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10050372
  • € 32.95 / Pack of 25
Estimated Shipment: 26-06-2023
to see stock.
Add to basket
Description
Description
Powder Pillows make titration applications quick and easy.
  • Sealed packets ensure fresh, uncontaminated reagent every time
  • Premeasured to avoid measurement error and contamination from handling
  • Ready to use packets save time and hassle
  • Exact doses to easily monitor supply and prepare for lab procedures
  • Single-use packs for easy transportation and packing in test kits
Specifications
Specifications
Zinc
Individual Packet
25
0.01 to 2.00mg/L Zn
Zincon
US EPA
Zinc determination
25 Pack
50 mL
Reagent packet