Learn More
ZIP13 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26496-1-AP-150UL
 View more versions of this product
Product Code. 18024475
  • € 410.00 / 150µL
Description
The ZIP13 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ZIP13. This antibody recognizes human, mouse, rat antigen. The ZIP13 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
ZIP13
1000 μg/mL
Rabbit
Primary
Human, Mouse, Rat
150 kDa
IgG
Western Blot, ELISA
LZT Hs9, SLC39A13, Zinc transporter ZIP13, ZIP 13, ZIP13, Zrt and Irt like protein 13
150 μL
91252
Store at -20°C. Stable for one year after shipment.