Learn More
ZIP13 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26496-1-AP-150UL
Description
The ZIP13 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ZIP13. This antibody recognizes human, mouse, rat antigen. The ZIP13 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| ZIP13 | |
| 1000 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| LZT Hs9, SLC39A13, Zinc transporter ZIP13, ZIP 13, ZIP13, Zrt and Irt like protein 13 | |
| 150 μL | |
| 91252 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |