Learn More
Invitrogen™ ZIP14 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5140224
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: STCFSSGDLF TAHNFSEQSR IGSSELQEFC PTILQQLDSR ACTSENQENE ENEQTEEGRP SAVEVW Highest antigen sequence indentity to the following orthologs: Mouse - 83%, Rat - 88%.
The zinc transporter ZIP14, also known as SLC39A14, is a member of a family of divalent ion transporters. Zinc is an essential ion for cells and plays significant roles in the growth, development, and differentiation. The zinc transporter family is divided into four subfamilies (I, II, LIV-1 and gufA). ZIP14 is a glycosylated multipass plasma membrane protein that belongs to the ZIP transporter subfamily LIV-1. ZIP14 has been shown to contribute to the hypozincemia of inflammation and infection and is regulated in the liver by IL-6. In addition to zinc, ZIP14 is also involved in the cellular uptake of non-transferrin-bound iron as well as iron bound to transferrin.
Specifications
| ZIP14 | |
| Polyclonal | |
| Unconjugated | |
| SLC39A14 | |
| cig19; Factor for adipocyte differentiation 123; Fad123; FAD-123; HMNDYT2; Kiaa0062; LIV-1 subfamily of ZIP zinc transporter 4; LZT-Hs4; NET34; SLC39A14; solute carrier family 39 (metal ion transporter), member 14; solute carrier family 39 (zinc transporter), member 14; solute carrier family 39 member 14; Zinc transporter ZIP14; Zip14; ZIP-14; Zrt- and Cig19; Zrt- and Irt-like protein 14; Zrt-, Irt-like protein 14 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23516 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15043 | |
| SLC39A14 | |
| Recombinant protein corresponding to Human SLC39A14. Recombinant protein control fragment (Product #RP-108930). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.