Learn More
Thermo Scientific™ ZipChip™ Assay Kits
Product Code. 17881691 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Intact Protein Assay Kit
Metabolites Assay Kit
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
17881691 Metabolites Assay Kit 1 set
17891691 Intact Protein Assay Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17881691 Supplier Thermo Scientific™ Supplier No. 009500100500
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Reduce error and enjoy the convenience of prepackaged ZipChip™ Assay Kits, which are for use on the ZipChip Interface for analysis of biological samples by MS.
Reduce error and enjoy the convenience of prepackaged ZipChip™ Assay Kits. These kits are for use with the ZipChip Interface, with or without the ZipChip Autosampler. Each ZipChip Assay Kit contains 5 bottles of background electrolyte and sample diluents, which is sufficient for 500 sample injections. A bottle can be directly loaded into ZipChip Autosampler for automated use and matching to a 96-well plate run. Different kits have been optimized for peptides, intact antibody, and metabolites analyses.
Three types of pre-packed ZipChip Assay Kits are customized for analyses of intact proteins, pepetides, and metabolites
  • Total of 500-sample capacity per kit
  • 5 individual bottles with 100 sample capacity each (designed to match to 96-well plate sampling) can be directly loaded into ZipChip autosampler
Specifications
Type Metabolites Assay Kit
For Use With (Equipment) ZipChip Interface for Mass Spectrometry
Includes Background electrolyte and sample diluents optimized for metabolite analysis