Learn More
Thermo Scientific Chemicals Zirconium Oxide Round Bottom Crucibles, High Form
Click to view available options
Capacity (Metric):
5 mL
10 mL
100 mL
200 mL
Diameter (Metric) Outer:
26mm
30mm
56mm
69mm
Height (Metric):
29mm
34mm
68mm
85mm
Unit Size:
Each
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Especificaciones
| Material | Zirconium Oxide |
| Quantity | 1 Piece |
| Capacity (Metric) | 200 mL |
| Diameter (Metric) Outer | 69mm |
| Height (Metric) | 85mm |
| Product Type | Zirconium Oxide Round Bottom Crucibles, High Form |
| Diameter (Metric) Base | 42mm |
RUO – Research Use Only
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido