Learn More
Thermo Scientific™ Zirconium Oxide Round Bottom Crucibles, Low Form
Product Code. 15423845
missing translation for 'orderingAttributeHoverText'
Capacity (Metric):
5 mL
100 mL
200 mL
Diameter (Metric) Outer:
29mm
67mm
82mm
Height (Metric):
21mm
51mm
63mm
missing translation for 'unitSize'
Each
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15423845
missing translation for 'mfr': Thermo Scientific™ 041296.KF
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Material Zirconium Oxide
Quantity 1 Piece
Capacity (Metric) 200 mL
Diameter (Metric) Outer 82mm
Height (Metric) 63mm
Product Type Zirconium Oxide Round Bottom Crucibles, Low Form
Diameter (Metric) Base 42mm