missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZKSCAN3 Antibody (2A1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00080317-M09
This item is not returnable.
View return policy
Description
ZKSCAN3 Monoclonal antibody specifically detects ZKSCAN3 in Human samples. It is validated for Western Blot, ELISA
Specifications
| ZKSCAN3 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| dJ874C20.1, FLJ33906, ZF47, Zfp47, Zfp-47, Zinc finger and SCAN domain-containing protein 13, Zinc finger protein 306KIAA0426, Zinc finger protein 309ZNF306ZFP306, Zinc finger protein 47 homolog, zinc finger protein with KRAB and SCAN domains 3, zinc finger protein zfp47, zinc finger with KRAB and SCAN domains 3, ZNF309, ZSCAN13 | |
| ZNF306 (NP_077819, 431 a.a. ∽ 536 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. CGKAFRRSSHLLRHQRIHTGDKNVQEPEQGEAWKSRMESQLENVETPMSYKCNECERSFTQNTGLIEHQKIHTGEKPYQCNACGKGFTRISYLVQHQRSHVGKNIL | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 2A1 | |
| Western Blot 1:500, ELISA | |
| NP_077819 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 80317 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction