missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZKSCAN4 Monoclonal Antibody (OTI6G6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800605S
This item is not returnable.
View return policy
Description
ZKSCAN4 Monoclonal antibody specifically detects ZKSCAN4 in Human samples. It is validated for Western BlotSpecifications
| ZKSCAN4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P05412 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 3725 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI6G6 | |
| Unconjugated | |
| JUN | |
| AI841875; Gm11272; OTTMUSG00000000421; P1P373C6; P373c6.1; p373c6.1 (novel C2H2 type zinc finger protein); Zfp307; Zinc finger protein 307; zinc finger protein 427; Zinc finger protein with KRAB and SCAN domains 4; zinc finger with KRAB and SCAN domains 4; ZKSCAN4; ZNF307; ZNF427; ZSCAN36 | |
| Full length human recombinant protein of human JUN produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction