missing translation for 'onlineSavingsMsg'
Learn More

ZLN024 hydrochloride, MedChemExpress

Product Code. 16497148
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497148 10 mg 10mg
16463177 50 mg 50mg
16429687 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16497148 Supplier Medchem Express Supplier No. HY16708A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ZLN024 hydrochloride is an AMPK allosteric activator. ZLN024 directly activates recombinant AMPK α1β1γ1, AMPK α2β1γ1, AMPK α1β2γ1 and AMPK α2β2γ1 heterotrimer with EC50s of 0.42 μM, 0.95 μM, 1.1 μM and 0.13 μM, respectively.

ZLN024 hydrochloride is an AMPK allosteric activator. ZLN024 directly activates recombinant AMPK α1β1γ1, AMPK α2β1γ1, AMPK α1β2γ1 and AMPK α2β2γ1 heterotrimer with EC50s of 0.42 μM, 0.95 μM, 1.1 μM and 0.13 μM, respectively.

Specifications

Chemical Name or Material ZLN024 hydrochloride
CAS 1883548-91-1
Color White
Purity Grade Notes Research
Molecular Formula C13H14BrClN2OS
Quantity 10 mg
Solubility Information DMSO : ≥ 46 mg/mL (127.18 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES CC1=CC=C(OCCSC2=NC=CC=N2)C(Br)=C1.[H]Cl
Molecular Weight (g/mol) 361.69
Formula Weight 361.69
Percent Purity 98.54%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.