missing translation for 'onlineSavingsMsg'
Learn More

ZM 306416, MedChemExpress

Product Code. p-200097435
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16482007

Brand: Medchem Express HY13785

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ZM-306416 (CB 676475) is a potent inhibitor of VEGFR with IC50s of 0.1 and 2 μM for KDR and Flt, respectively. ZM-306416 is also a EGFR inhibitor with an IC50 of <10 nM.

ZM-306416 (CB 676475) is a potent inhibitor of VEGFR with IC50s of 0.1 and 2 μM for KDR and Flt, respectively. ZM-306416 is also a EGFR inhibitor with an IC50 of <10 nM.

Specifications

Chemical Name or Material ZM 306416
CAS 690206-97-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C16H13ClFN3O2
Quantity 50 mg
Synonym CB 676475
Solubility Information DMSO : 50 mg/mL (149.82 mM; Need ultrasonic)
SMILES COC1=CC2=NC=NC(NC3=C(F)C=C(Cl)C=C3)=C2C=C1OC
Molecular Weight (g/mol) 333.74
Formula Weight 333.74
Percent Purity 99.02%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.