missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZMAT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5118369
This item is not returnable.
View return policy
Description
ZMAT1 Polyclonal Antibody for Western Blot, IHC (P), IP
This gene encodes a protein containing Cys2-His2 (C2H2)-type zinc fingers, which are similar to those found in the nuclear matrix protein matrin 3. Alternatively spliced transcript variants have been found for this gene. [provided by RefSeq, Jan 2012].
Specifications
| ZMAT1 | |
| Polyclonal | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q5H9K5 | |
| ZMAT1 | |
| E. coli-derived Human ZMAT1 fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| ZMAT1 | |
| A230096A06; AW990744; B930008K04Rik; KIAA1789; RGD1565344; zinc finger matrin-type 1; zinc finger matrin-type 1; zinc finger matrin-type protein 1; zinc finger matrin-type protein 1; zinc finger, matrin type 1; zinc finger, matrin-type 1; ZMAT1 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 84460 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction