missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZMAT4 Antibody (2A4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00079698-M02
This item is not returnable.
View return policy
Description
ZMAT4 Monoclonal antibody specifically detects ZMAT4 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
| ZMAT4 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| FLJ13842, zinc finger matrin-type protein 4, zinc finger, matrin type 4, zinc finger, matrin-type 4 | |
| ZMAT4 (AAH19598, 1 a.a. ∽ 153 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MKSSDIDQDLFTDSYCKVCSAQLISESQRVAHYESRKHASKVRLYYMLHPRDGGCPAKRLRSENGSDADMVDKNKCCTLCNMSFTSAVVADSHYQGKIHAKRLKLLLGEKTPLKTTGLRRNYRCAICSVSLNSIEQYHAHLKGSKHQTNLKNK | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunoprecipitation | |
| 2A4 | |
| Western Blot 1:500, ELISA, Immunoprecipitation | |
| AAH19598 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 79698 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction