missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZMAT4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals H00079698-D01P
This item is not returnable.
View return policy
Description
ZMAT4 Polyclonal antibody specifically detects ZMAT4 in Human samples. It is validated for Western Blot
Specifications
| ZMAT4 | |
| Polyclonal | |
| Western Blot 1:500 | |
| AAH19598.1 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 79698 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.4) | |
| FLJ13842, zinc finger matrin-type protein 4, zinc finger, matrin type 4, zinc finger, matrin-type 4 | |
| ZMAT4 (AAH19598.1, 1 a.a. - 153 a.a.) full-length human protein. MKSSDIDQDLFTDSYCKVCSAQLISESQRVAHYESRKHASKVRLYYMLHPRDGGCPAKRLRSENGSDADMVDKNKCCTLCNMSFTSAVVADSHYQGKIHAKRLKLLLGEKTPLKTTGLRRNYRCAICSVSLNSIEQYHAHLKGSKHQTNLKNK | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction