missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZMAT4 Monoclonal Antibody (OTI9E11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809685
This item is not returnable.
View return policy
Description
ZMAT4 (Zinc Finger Matrin-Type 4) is a Protein Coding gene. Diseases associated with ZMAT4 include Refractive Error. Gene Ontology (GO) annotations related to this gene include nucleic acid binding. An important paralog of this gene is ZNF346.Specifications
| ZMAT4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9H898 | |
| ZMAT4 | |
| Full length human recombit protein of human ZMAT4 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI9E11 | |
| Unconjugated | |
| ZMAT4 | |
| zinc finger matrin-type 4 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 79698 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction