missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZMIZ2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93245-0.02ml
This item is not returnable.
View return policy
Description
ZMIZ2 Polyclonal antibody specifically detects ZMIZ2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| ZMIZ2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| hZIMP7, KIAA1886ZIMP7DKFZp761I2123, NET27, PIAS-like protein Zimp7, TRAFIP20, zinc finger MIZ domain-containing protein 2, zinc finger, MIZ-type containing 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-110 of human ZMIZ2 (NP_001287888). MNSMNPMKPALPPAPHGDGSFAYESVPWQQSATQPAGSLSVVTTVWGVGNATQSQCLGQQAFAEGGANKGYVQQGVYSRGGYPGAPGFTTGYAGGPGGLGLPSHAARPST | |
| 0.02 mL | |
| Chromatin Modifiers, Signal Transduction | |
| 83637 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction