missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZMYM1 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76523PCP
This item is not returnable.
View return policy
Description
ZMYM1 Polyclonal antibody specifically detects ZMYM1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| ZMYM1 | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| FLJ23151, zinc finger, MYM domain containing 1, zinc finger, MYM-type 1 | |
| Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human ZMYM1 . The immunogen is located within amino acids 990 - 1040 of ZMYM1. Amino Acid Squence: WNEPLNETTAKHVQEFY | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| PerCP | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 79830 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction