missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZMYM1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566658
This item is not returnable.
View return policy
Description
Immunogen sequence: NSTQIQSDIIE IIKTEMLQDI VNEINDSSAF SIICDETINS AMKEQLSICV RYPQKSSKAI LIKERFLGFV DTEEMTGTHL Highest antigen sequence identity to the following orthologs - mouse 67%, rat 64%.
Zinc-finger proteins contain DNA-binding domains characterized by the unique role of zinc and have a wide variety of functions such as transcriptional activation or repression. The protein folding and the DNA binding ability are governed by the coordination of a zinc ion. As a member of the MYM (myeloproliferative and mental retardation) gene family, ZMYM1 is widely expressed in different tissues in eukaryotes under several forms derived by alternative splicing. While its function remains unknown, the related protein ZMYM2 has been shown to associate with and stabilize the LSD1-CoREST-HDAC1 (LCH) complex of chromatin through its MYM-type zinc fingers, thereby enhancing the transcriptional repression of several genes, suggesting that ZMYM1 may play a similar role.
Specifications
| ZMYM1 | |
| Polyclonal | |
| Unconjugated | |
| ZMYM1 | |
| 5830412B09Rik; AW209050; MYM; RP11-181E22.4; RP11-248I9.4; zinc finger MYM-type containing 1; zinc finger MYM-type protein 1; zinc finger, MYM domain containing 1; zinc finger, MYM-type 1; ZMYM1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79830 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5SVZ6 | |
| ZMYM1 | |
| Recombinant Human ZMYM1. Recombinant protein control fragment (Product #RP-107306). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction