missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZMYM3 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98112UV
This item is not returnable.
View return policy
Description
ZMYM3 Polyclonal specifically detects ZMYM3 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| ZMYM3 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| DXS6673EZinc finger protein 261KIAA0385XFIM, MYM, zinc finger, MYM-type 3, ZNF198L2, ZNF261zinc finger MYM-type protein 3 | |
| Produced in rabbits immunized with E. coli-derived Human ZMYM3 fragment. | |
| 0.1 mL | |
| DNA Repair | |
| 9203 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunofluorescence | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction