missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZMYM3 Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-458V
This item is not returnable.
View return policy
Description
ZMYM3 Polyclonal antibody specifically detects ZMYM3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| ZMYM3 | |
| Polyclonal | |
| Rabbit | |
| Immunogen affinity purified | |
| Primary | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| DyLight 405 | |
| The immunogen this antibody was made to, maps to a region between residues 250-350 of human Zinc Finger Protein 261 using the NP_005087.1 entry Q8IY18 (GeneID 9203). | |
| 0.1 mL | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction