missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZMYM3 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA551862

Product Code. 15709820

  • € 578.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: PEVDHGPEGT LAWDAGDQTL EPGPGGQTPE VVPPDPGAGA NSCSPEGLLE PLAPDSPITL QSPHIEEEET TSIATARRGS PGQEEELPQG QPQSPNAPPS PSVGETLGDG INSSQTKPGG SSPPAHPSLP GDGLTAKASE KPPERKRS Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 83%.

This gene is located on the X chromosome and is subject to X inactivation. It is highly conserved in vertebrates and most abundantly expressed in the brain. The encoded protein is a component of histone deacetylase-containing multiprotein complexes that function through modifying chromatin structure to keep genes silent. A chromosomal translocation (X;13) involving this gene is associated with X-linked cognitive disability. Several alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ZMYM3
Polyclonal
Unconjugated
ZMYM3
9030216B10Rik; AW122925; DXHXS6673E protein; DXS6673E; DXS6673El; KIAA0385; MYM; XFIM; Zfp261; zinc finger MYM-type containing 3; zinc finger MYM-type protein 3; Zinc finger protein 261; zinc finger, MYM-type 3; Zmym3; ZNF198L2; ZNF261
Rabbit
Antigen affinity chromatography
RUO
9203
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry
0.6 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q14202
ZMYM3
Recombinant protein corresponding to Human ZMYM3. Recombinant protein control fragment (Product #RP-92213).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.