missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZMYM4 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-95000IR
This item is not returnable.
View return policy
Description
ZMYM4 Polyclonal antibody specifically detects ZMYM4 in Human samples. It is validated for Western Blot
Specifications
| ZMYM4 | |
| Polyclonal | |
| Western Blot | |
| CDIR, cell death inhibiting RNA, KIAA0425DKFZp686L0547, MYM, zinc finger MYM-type protein 4, Zinc finger protein 262DKFZp686B09210, zinc finger, MYM-type 4, ZNF198L3, ZNF262 | |
| The immunogen recognized by this antibody maps to a region between residue 850 and 900 of human zinc finger, MYM-type 4 using the numbering given in entry NP_005086.2 (GeneID 9202). | |
| 0.1 mL | |
| Cell Biology | |
| 9202 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction