Learn More
Invitrogen™ ZMYM4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589455
Description
Immunogen sequence: TEMSEDIDHN LTPTLDSMSY GMPNQTGSEN SLLDEDDYFL NSGDLAGIPV VGSDNEDEQD FSSKDNLVSS IHTDDSLEVE RRVTQHESDN ENEIQIQNKL KKDFPKQFDQ VSVFKSIRKD FSLVRENSKE TFSGKEKNRD LTYEREKRLD KPHKDLDSRL KSSFFDKAAN QVEETLHTHL PQTPETNFRD SSYPFA; Positive Samples: 293T, HepG2, U87, HeLa.
Zinc-finger proteins contain DNA-binding domains characterized by the unique role of zinc and have a wide variety of functions such as transcriptional activation or repression. The protein folding and the DNA binding ability are governed by the coordination of a zinc ion. It has been found to be overexpressed in human lung adenocarcinomas and squamous cell carcinomas, and the overexpression of a fragment of the 3'UTR of the ZMYM4 mRNA termed Cell Death Inhibiting RNA (CDIR) protects HeLa cells from IFN-gamma-induced apoptosis, suggesting that ZMYM4 may play a role in tumorigenesis.
Specifications
| ZMYM4 | |
| Polyclonal | |
| Unconjugated | |
| ZMYM4 | |
| 6330503C17Rik; AI480785; AW493829; CDIR; cell death inhibiting RNA; D630001M21; Kiaa0425; mKIAA0425; MYM; MYM type 4; Zfp262; zinc finger MYM-type containing 4; zinc finger MYM-type protein 4; Zinc finger protein 262; zinc finger, MYM-type 4; Zmym4; ZNF198L3; ZNF262 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 9202 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.44 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q5VZL5 | |
| ZMYM4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 35-230 of human ZMYM4 (NP_0050862). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.