missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZMYM6 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-26106H
This item is not returnable.
View return policy
Description
ZMYM6 Polyclonal antibody specifically detects ZMYM6 in Human samples. It is validated for Western Blot
Specifications
| ZMYM6 | |
| Polyclonal | |
| Western Blot | |
| Buster2, KIAA1353, MYM, MYM-type containing 6, zinc finger MYM-type protein 6, zinc finger, MYM-type 6 | |
| Produced in rabbits immunized with E. coli-derived Human ZMYM6 fragment | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| HRP | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 9204 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction