missing translation for 'onlineSavingsMsg'
Learn More

ZNF10 Antibody (CL9744) [Alexa Fluor« 750], Novus Biologicals Biologicals™

Product Code. 30529743 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30529743 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30529743 Supplier Novus Biologicals Supplier No. NBP344232AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZNF10 Monoclonal antibody specifically detects ZNF10 in Human samples. It is validated for Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF10
Applications Western Blot, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone CL9744
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias KOX1zinc finger protein 10 (KOX 1), zinc finger protein 10, Zinc finger protein KOX1
Host Species Mouse
Immunogen This antibody was generated using a synthetic peptide of P21506, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 7556
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.