Learn More
ZNF10 Antibody (CL9744) [Janelia Fluor™ 549], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-88939JF549
Product Code. 30293265
  • € 737.00 / 0.10mL
Description
ZNF10 Monoclonal antibody specifically detects ZNF10 in Human samples. It is validated for Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ZNF10
Monoclonal
Janelia Fluor 549
KOX1zinc finger protein 10 (KOX 1), zinc finger protein 10, Zinc finger protein KOX1
This antibody was developed using a synthetic peptide derived from P21506, with the exact immunogen sequence remaining proprietary.
0.1 mL
Transcription Factors and Regulators
7556
Store at 4⪚C in the dark.
IgG1
Western Blot, Immunocytochemistry/Immunofluorescence
CL9744
50mM Sodium Borate
Mouse
Protein A purified