missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF100 Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97820AF700
This item is not returnable.
View return policy
Description
ZNF100 Polyclonal specifically detects ZNF100 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| ZNF100 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 163227 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 700 | |
| FLJ44587, zinc finger protein 100, zinc finger protein 100 (Y1) | |
| Produced in rabbits immunized with E. coli-derived Human ZNF100 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction