missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF114 Antibody (OTI6F4), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74931AF700
This item is not returnable.
View return policy
Description
ZNF114 Monoclonal specifically detects ZNF114 in Human samples. It is validated for Western Blot.
Specifications
| ZNF114 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ZNF114 | |
| Full length recombinant protein of human ZNF114 (NP_705836) produced in E.coli. | |
| 0.1 mL | |
| 163071.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI6F4 | |
| Western Blot | |
| MGC149700, MGC17986, zinc finger protein 114, zinc finger protein 114 (Y18) | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction