missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF140 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97821AF750
This item is not returnable.
View return policy
Description
ZNF140 Polyclonal specifically detects ZNF140 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| ZNF140 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| EC 1.3.1.26, EC 2.1.1.43, pHZ-39, zinc finger protein 140, zinc finger protein 140 (clone pHZ-39) | |
| Produced in rabbits immunized with E. coli-derived Human ZNF140 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 750 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 7699 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction