Learn More
ZNF141 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-05597-100ul
 View more versions of this product
Product Code. 18622889
  • € 463.00 / 100µL
Description
ZNF141 Polyclonal antibody specifically detects ZNF141 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ZNF141
Polyclonal
Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin
D4S90, pHZ-44, zinc finger protein 141
Recombinant fusion protein containing a sequence corresponding to amino acids 405-474 of human ZNF141 (NP_003432.1). RRSTDRSQHKKIHSADKPYKCKECDKAFKQFSLLSQHKKIHTVDKPYKCKDCDKAFKRFSHLNKHKKIHT
100 μg
Primary
Human, Mouse, Rat
Purified
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
PBS with 50% glycerol, pH7.3.
Rabbit
Affinity purified
RUO
7700
Store at -20°C. Avoid freeze-thaw cycles.