missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF148 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-71872FR
This item is not returnable.
View return policy
Description
ZNF148 Polyclonal antibody specifically detects ZNF148 in Human, Mouse samples. It is validated for Western Blot
Specifications
| ZNF148 | |
| Polyclonal | |
| Western Blot | |
| BERF-1, BFCOL1, CACCC box-binding protein, CLL-associated antigen KW-10, HT-BETA, pHZ-52, Transcription factor ZBP-89, ZBP89, ZBP-89, ZFP148zinc finger protein 148 (pHZ-52), Zinc finger DNA-binding protein 89, zinc finger protein 148 | |
| The immunogen for this product maps to a region between residue 744 and 794 of human Zinc Finger DNA-Binding Protein 89 using the numbering given in entry NP_068799.2 (GeneID 7707). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 7707 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction