missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF154 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22161-1-AP-20UL
This item is not returnable.
View return policy
Description
ZNF154 may be involved in transcriptional regulation.Specifications
| ZNF154 | |
| Polyclonal | |
| Unconjugated | |
| ZNF154 | |
| KIAA2003, pHZ 92, zinc finger protein 154, ZNF154 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 7710 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q13106 | |
| ZNF154 | |
| ZNF154 Fusion Protein Ag17812 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction