missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF157 Antibody (PCRP-ZNF157-1A8), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24002AF750
This item is not returnable.
View return policy
Description
ZNF157 Monoclonal antibody specifically detects ZNF157 in Human samples. It is validated for Flow Cytometry
Specifications
| ZNF157 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| HZF22, zinc finger protein 157, zinc finger protein 157 (HZF22), zinc finger protein 22, Zinc finger protein HZF22 | |
| Protein domain of ZNF157 protein | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| PCRP-ZNF157-1A8 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 7712 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction