missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF165 Antibody (OTI4C1), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74932B
This item is not returnable.
View return policy
Description
ZNF165 Monoclonal specifically detects ZNF165 in Human samples. It is validated for Western Blot.
Specifications
| ZNF165 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| ZNF165 | |
| Recombinant protein fragment corresponding to amino acids 165-485 of human ZNF165 (NP_003438) produced in E.coli. | |
| 0.1 mL | |
| 7718.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI4C1 | |
| Western Blot | |
| Cancer/testis antigen 53, CT53Zinc finger and SCAN domain-containing protein 7, zinc finger protein 165, ZSCAN7LD65ZPF165 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction