missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF18 Monoclonal Antibody (OTI2C1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810147S
This item is not returnable.
View return policy
Description
ZNF18 Monoclonal antibody specifically detects ZNF18 in Human samples. It is validated for Western BlotSpecifications
| ZNF18 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BSV6 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 79042 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI2C1 | |
| Unconjugated | |
| TSEN34 | |
| 1700128E15Rik; D11Ertd714e; HDSG1; Heart development-specific gene 1 protein; KOX11; Zfp18; Zfp535; Zinc finger protein 18; zinc finger protein 18 (KOX 11); zinc finger protein 535; Zinc finger protein KOX11; Zinc finger protein with KRAB and SCAN domains 6; zinc finger with KRAB and SCAN domains 6; Zkscan6; Znf18; ZNF535; ZSCAN38 | |
| Full length human recombinant protein of human TSEN34 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction