missing translation for 'onlineSavingsMsg'
Learn More

ZNF18 Monoclonal Antibody (OTI4C5), TrueMAB™, OriGene

Product Code. p-200015741
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17542924 30 μL 30µL
17522924 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17542924 Supplier Origene Technologies Supplier No. TA810108S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZNF18 Monoclonal antibody specifically detects ZNF18 in Human samples. It is validated for Western Blot

Specifications

Antigen ZNF18
Applications Western Blot
Classification Monoclonal
Clone OTI4C5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene KRT18P55
Gene Alias 1700128E15Rik; D11Ertd714e; HDSG1; Heart development-specific gene 1 protein; KOX11; Zfp18; Zfp535; Zinc finger protein 18; zinc finger protein 18 (KOX 11); zinc finger protein 535; Zinc finger protein KOX11; Zinc finger protein with KRAB and SCAN domains 6; zinc finger with KRAB and SCAN domains 6; Zkscan6; Znf18; ZNF535; ZSCAN38
Host Species Mouse
Immunogen Full length human recombinant protein of human FLJ40504 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284085
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.