missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF181 Antibody (5F1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00339318-M01
This item is not returnable.
View return policy
Description
ZNF181 Monoclonal antibody specifically detects ZNF181 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| ZNF181 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| zinc finger protein 181 | |
| ZNF181 (XP_290835, 354 a.a. ∽ 461 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. FIHRSSLIHHQKIHTGEKPYECRECGKAFCCSSHLTRHQRIHTMEKQYECNKCLKVFSSLSFLVQHQSIHTEEKPFECQKCRKSFNQLESLNMHLRNHIRLKPYECSI | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| 5F1 | |
| Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence | |
| XP_290835.3 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 339318 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction