missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF182 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584425
This item is not returnable.
View return policy
Description
Immunogen sequence: YSNLVFVGQQ VTKPNLILKL EVEECPAEGK IPFWNFPEVC QVDE Highest antigen sequence indentity to the following orthologs: Mouse - 62%, Rat - 51%.
Zinc-finger proteins bind nucleic acids and play important roles in various cellular functions, including cell proliferation, differentiation, and apoptosis. This gene encodes a zinc finger protein, and belongs to the krueppel C2H2-type zinc-finger protein family. It may be involved in transcriptional regulation. Multiple alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
| ZNF182 | |
| Polyclonal | |
| Unconjugated | |
| ZNF182 | |
| 6330583I20Rik; HHZ150; KOX14; MGC137263 protein; Zfp182; Zinc finger protein 182; zinc finger protein 21; zinc finger protein 21 (KOX 14); zinc finger protein KOX14; ZNF182; ZNF21 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7569 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P17025 | |
| ZNF182 | |
| Recombinant protein corresponding to Human ZNF182. Recombinant protein control fragment (Product #RP-101936). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction