missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF192 Monoclonal Antibody (3B10)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA520289
This item is not returnable.
View return policy
Description
Peptide Sequence: MAEESRKPSA PSPPDQTPEE DLVIVKVEED HGWDQESSLH ESNPLGQEVF RLRFRQLRYQ ETLGPREALI QLRALCHQWL RPDLNTKEQI.
May be involved in transcriptional regulation. [UniProt].
Specifications
| ZNF192 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q15776 | |
| ZKSCAN8 | |
| ZKSCAN8 (NP_006289.2, 1 a.a. ~ 90 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| 3B10 | |
| Unconjugated | |
| ZKSCAN8 | |
| 2510038J07Rik; CAML; D430019P06Rik; LD5-1; Zfp192; zinc finger protein 192; Zinc finger protein with KRAB and SCAN domains 8; zinc finger with KRAB and SCAN domains 8; ZKSCAN8; ZNF192; ZSCAN40 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 7745 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction