missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF198 Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-56490MFV500
This item is not returnable.
View return policy
Description
ZNF198 Polyclonal antibody specifically detects ZNF198 in Human, Canine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| ZNF198 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| FIMRAMPSCLL, Fused in myeloproliferative disorders protein, MYM, Rearranged in atypical myeloproliferative disorder protein, Zinc finger protein 198rearranged in an atypical myeloproliferative disorder, zinc finger, MYM-type 2, ZNF198zinc finger MYM-type protein 2 | |
| A portion of amino acids 1250-1300 of human Zinc finger protein 198 was used as the immunogen. | |
| 0.1 mL | |
| Primary | |
| Human, Canine, Primate | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 7750 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction