missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF198 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 241.00 - € 326.00
Specifications
| Antigen | ZNF198 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:1000-1:4000 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18647822
|
Novus Biologicals
NBP2-95284-100ul |
100 μg |
€ 326.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18684515
|
Novus Biologicals
NBP2-95284-50ul |
50 μg |
€ 241.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZNF198 Polyclonal antibody specifically detects ZNF198 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| ZNF198 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| FIMRAMPSCLL, Fused in myeloproliferative disorders protein, MYM, Rearranged in atypical myeloproliferative disorder protein, Zinc finger protein 198rearranged in an atypical myeloproliferative disorder, zinc finger, MYM-type 2, ZNF198zinc finger MYM-type protein 2 | |
| Produced in rabbits immunized with A synthetic peptide corresponding to the C-terminus of the human ZNF198. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry-Paraffin 1:1000-1:4000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 7750 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title