missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF20 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93053-0.1ml
This item is not returnable.
View return policy
Description
ZNF20 Polyclonal antibody specifically detects ZNF20 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| ZNF20 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| FLJ39241, zinc finger protein 20, zinc finger protein KOX13 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human ZNF20 (NP_066966.2). MMFQDSVAFEDVAVSFTQEEWALLDPSQKNLYRDVMQETFKNLTSVGKTWKVQNIEDEYKNPRRNLSLMREKLCESKESHHCGESFNQIADDMLNRKTLPGITPCESSVCGEVGTGHSSLNTHIRADTGHKSSEYQEYGENPYRNKECKKAFSYLDSFQSHDKACTKEKPYDGKECTETFISHSCIQRHRVMHSGDGPYK | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 7568 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction