missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF202 Antibody (PCRP-ZNF202-1C4), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24003JF549
This item is not returnable.
View return policy
Description
ZNF202 Monoclonal antibody specifically detects ZNF202 in Human samples. It is validated for Flow Cytometry
Specifications
| ZNF202 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| zinc finger protein 202, ZKSCAN10Zinc finger protein with KRAB and SCAN domains 10 | |
| Recombinant fragment (around 37-132) of human ZNF202 protein | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| PCRP-ZNF202-1C4 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 7753 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction