Learn More
ZNF202 Mouse anti-Human, Clone: OTI1D12, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF811039
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
ZNF202 (Zinc finger protein 202) is a transcriptional repressor of genes affecting the vascular endothelium as well as lipid metabolism and energy homeostasis. Among its targets are structural components of lipoprotein particles (apolipoproteins AIV, CIII, and E), enzymes involved in lipid processing (lipoprotein lipase, lecithin cholesteryl ester transferase), transporters involved in lipid homeostasis (ABCA1, ABCG1), and several genes involved in processes related to energy metabolism and vascular disease.Specifications
| ZNF202 | |
| Monoclonal | |
| Unconjugated | |
| ZNF202 | |
| ZKSCAN10, ZSCAN42 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 7753 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI1D12 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| O95125 | |
| ZNF202 | |
| Human recombit protein fragment corresponding to amino acids 1-307 of human ZNF202 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.