missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF205 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-06486CL1
This item is not returnable.
View return policy
Description
ZNF205 Polyclonal antibody specifically detects ZNF205 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence
Specifications
| ZNF205 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 7755 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| zinc finger protein 205, Zinc finger protein 210, ZNF210Zfp13 | |
| Produced in rabbits immunized with E. coli-derived Human ZNF205 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction